missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSTPIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11951-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene binds to the cytoplasmic tail of CD2, an effector of T cell activation and adhesion, negatively affecting CD2-triggered T cell activation. The encoded protein appears to be a scaffold protein and a regulator of the actin cytoskeleton. It has also been shown to bind ABL1, PTPN18, WAS, CD2AP, and PTPN12. Mutations in this gene are a cause of PAPA syndrome.Specifications
| PSTPIP1 | |
| Polyclonal | |
| Unconjugated | |
| PSTPIP1 | |
| CD2 binding protein 1, CD2BP1, CD2BP1L, CD2BP1S, H PIP, PAPAS, PSTPIP, PSTPIP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19200, 300732, 9051 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43586, P97814 | |
| Pstpip1 | |
| PSTPIP1 Fusion Protein Ag2551 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction