Learn More
Testo™ Pt100 Robust Air Probe
Product Code. 11361114
Change view
Click to view available options
Temperature Range (Metric):
-50°C to +400°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
11361114 -50°C to +400°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Product Code. 11361114 Supplier Testo™ Supplier No. 0609 1773
  • ONLINE EXCLUSIVE PRICE
    197.40€
    103.00€ / Pack of 1
    Save 94.40€
    47% Off
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
R\u00E9cupel-bijdrage uitgesloten
Specifications
Type Probe, Robust Air
Certifications/Compliance 60751 Standard
Response Time 70 sec.
Temperature Range (Metric) -50°C to +400°C