missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTAFR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525329
This item is not returnable.
View return policy
Description
PTAFR Polyclonal Antibody for Western Blot
PTAFR shows structural characteristics of the rhodopsin gene family and binds platelet-activating factor (PAF). PAF is a phospholipid (1-0-alkyl-2-acetyl-sn-glycero-3-phosphorylcholine) that has been implicated as a mediator in diverse pathologic processes, such as allergy, asthma, septic shock, arterial thrombosis, and inflammatory processes.
Specifications
| PTAFR | |
| Polyclonal | |
| Unconjugated | |
| PTAFR | |
| PAF receptor; PAFR; PAF-R; platelet activating factor receptor; platelet-activating factor receptor; Ptafr | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 19204, 5724 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P25105, Q62035 | |
| PTAFR | |
| KLH conjugated synthetic peptide between 286-312 amino acids from the C-terminal region of human PTAFR. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction