Learn More
Masterflex™ PTFE 2-Way Direct Lift Solenoid Valve
Product Code. 11759759
Change view
Click to view available options
Material:
Teflon
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
11759759 Teflon Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11759759 Supplier Masterflex™ Supplier No. 0154009
  • ONLINE EXCLUSIVE PRICE
    323.00€
    217.07€ / Pack of 1
    Save 105.93€
    32% Off
Estimated Shipment: 10-03-2027
Specifications
For Use With (Application) Ideal for use with high-purity or corrosive fluids
Max. Operating Temperature (Metric) 70°C
Material Teflon
Quantity 1 Each
Type 2-way Direct Lift Solenoid Valve
Voltage 12 VDC