Learn More
Fisherbrand™ Dipper Beaker Shafts
Product Code. 15132569 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Dimensions (L x W):
12 x 12 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15132569 12 x 12 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15132569
Estimated Shipment: 15-10-2026
Add to basket
Add to basket
Suitable for sampling the most aggressive media. Fisherbrand™ Dipper Beaker Shafts are fully encapsulated in PTFE plastic for rigidity.
  • Shafts have a steel core
  • Totally chemically resistant and inert
  • Additional shafts available to extend the dippers
Specifications
Product Type Dipper Beaker
Material PTFE
Autoclavable Autoclavable
Closure Material PTFE
Color White
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
For Use With (Equipment) PTFE Dipper Beakers
Temperature (Metric) -200°C to +280°C
Dimensions (L x W) 12 x 12 mm
Dimensions (L x W x H) 12 x 12 x 600 mm
Length (Metric) Handle 600 mm
Width (Metric) 12 mm