Learn More
Horst™ PTFE HBSI Model Hard-Wearing Heating Tape
Product Code. 10769442
Change view
Click to view available options
Length (Metric):
0.8 m
1 m
1.5 m
1.7 m
2 m
3 m
4 m
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10769442 1.5 m Each
10456371 0.8 m Each
10200592 1.7 m Each
10436941 2 m Each
10097161 3 m Each
10620383 4 m Each
10749812 1 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10769442 Supplier Horst™ Supplier No. 20603
  • ONLINE EXCLUSIVE PRICE
    270.62€
    152.51€ / Pack of 1
    Save 118.11€
    43% Off
View all promotions
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Specifications
Dimensions (L x W) 1.5 Lm
Dimensions (L x W x H) 1.5 Lm
Length (Metric) 1.5 m
Electrical Requirements ∼230V Voltage
Model HBSI
Product Type Hard-wearing Heating Tape