Learn More
Horst™ PTFE HST Model Humidity-Proof Heating Cables
Product Code. 11943945
Change view
Click to view available options
Length (Metric) Power Cord:
1.1m
5.5m
6m
8m
9m
12m
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Power Cord Unit Size
11943945 12m Each
10619812 1.1m Each
10007171 5.5m Each
10532892 6m Each
10679422 8m Each
11933945 9m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11943945 Supplier Horst™ Supplier No. 20805
  • ONLINE EXCLUSIVE PRICE
    285.15€
    176.64€ / Pack of 1
    Save 108.51€
    38% Off
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Specifications
Dimensions (L x W) 12 Lm
Dimensions (L x W x H) 12 Lm
Electrical Requirements ∼230V Voltage
Model HST
Length (Metric) Power Cord 12m
Product Type Humidity-proof Heating Cable