Learn More
Poulten Graf™ Fortuna™ Optifix™ Basic Dispenser
Product Code. 10339800
Click to view available options
Volume (Metric):
0.4 to 2 mL
1 to 5 mL
10 to 50 mL
100 to 500 mL
2 to 10 mL
20 to 100 mL
40 to 200 mL
6 to 30 mL
60 to 300 mL
Unit Size:
Each
Product Code. 10339800
Brand: Poulten Graf™ 101 08058
Only null left
Add to basket
Add to basket
Poulten Graf™ Fortuna™ Optifix™ Basic Dispensers have a dispensing cylinder and piston made of glass.
For dosing of the most common chemicals, acids and crytallizing liquids (not suitable for solvents). Poulten Graf™ PTFE/PP/Fiberglass/Teflon/Ceramic Fortuna™ Optifix™ Basic Dispensers have a dispensing cylinder and piston made of glass.
  • Manufactured according to DIN EN ISO 9001:2008 using high quality materials
  • With solid PTFE encased dispensing piston made of glass
  • Cylinder coated with plastic to protect against breakage
  • With certificate, individually numbered
  • Ergonomic finger protection cap made of PP with volume setting system
  • Graduation on the scale is lasered and easy to read
  • Fast and easy volume setting with additional fine adjustment
Specifications
Product Type Fortuna Optifix Basic Dispenser
Color Clear
Material PTFE, Polypropylene, Fiberglass, Teflon, Ceramic
Volume (Metric) 100 to 500 mL
For Use With (Application) For dispensing aqueous solutions, acids and crystallizing liquids
Quantity Each