Learn More
- Home
- Products
- Hotplates and Stirrers
- Stirring Bars and Rods
- Fisherbrand™ PTFE Stir Bars with Pivot Ring
Fisherbrand™ PTFE Stir Bars with Pivot Ring
Product Code. 11838862 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Length (Metric):
8 mm
12 mm
13 mm
15 mm
20 mm
25 mm
30 mm
35 mm
40 mm
45 mm
50 mm
60 mm
70 mm
75 mm
Diameter (Metric):
3 mm
4.5 mm
6 mm
8 mm
10 mm
13 mm
Unit Size:
Each
Pack of 10
Pack of 5
27 product options available for selection
| Product Code. | Length (Metric) | Diameter (Metric) | Unit Size |
|---|---|---|---|
| 11838862 | 20 mm | 6 mm | Pack of 10 |
| 10018131 | 60 mm | 10 mm | Each |
| 11898862 | 60 mm | 10 mm | Pack of 10 |
| 11888962 | 13 mm | 8 mm | Pack of 10 |
| 12641156 | 50 mm | 8 mm | Pack of 5 |
| 11537812 | 45 mm | 8 mm | Pack of 5 |
| 11888862 | 45 mm | 8 mm | Pack of 10 |
| 12379507 | 15 mm | 4.5 mm | Pack of 5 |
| 16265811 | 70 mm | 10 mm | Pack of 10 |
| 10373592 | 70 mm | 10 mm | Each |
| 12661156 | 30 mm | 6 mm | Pack of 10 |
| 12359507 | 12 mm | 6 mm | Pack of 5 |
| 12651156 | 12 mm | 6 mm | Pack of 10 |
| 11517812 | 35 mm | 6 mm | Pack of 5 |
| 11868862 | 35 mm | 6 mm | Pack of 10 |
| 11868962 | 75 mm | 13 mm | Pack of 5 |
| 11527812 | 40 mm | 8 mm | Pack of 5 |
| 11878862 | 40 mm | 8 mm | Pack of 10 |
| 11808972 | 25 mm | 10 mm | Pack of 10 |
| 11507812 | 25 mm | 6 mm | Pack of 5 |
| 11848862 | 25 mm | 6 mm | Pack of 10 |
| 11597802 | 20 mm | 6 mm | Pack of 5 |
| 11898962 | 15 mm | 8 mm | Pack of 10 |
| 11587802 | 12 mm | 4.5 mm | Pack of 5 |
| 11818862 | 12 mm | 4.5 mm | Pack of 10 |
| 11808862 | 8 mm | 3 mm | Pack of 10 |
| 12349507 | 8 mm | 3 mm | Pack of 5 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
Artikelnummer.
11838862
Læg i indkøbskurv
10.04 EUR valid until 2026-12-31
BEST PRICE on Promo! Use promo code "28038" to get your promotional price.
BEST PRICE on Promo! Use promo code "28038" to get your promotional price.
Læg i indkøbskurv
missing translation for 'validMainframeMsg'
Tekniske data
Tekniske data
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Color | White |
| Description | PTFE encapsulated Alnico V magnet core; Cylindrical with Pivot Ring |
| For Use With (Equipment) | Containers & Vessels / Stirrer Hot Plates |
| Diameter (Metric) | 6 mm |
| Length (Metric) | 20 mm |
| Shape | Cylinder, Pivot ring |
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?