Learn More
Bohlender™ PTFE T-shaped Bore 3-Way Stopcock
Product Code. 10043511
Change view
Click to view available options
Thread Size:
GL14
GL18
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size Unit Size
10043511 GL18 Each
10043441 GL14 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10043511 Supplier Bohlender™ Supplier No. E 68818
  • ONLINE EXCLUSIVE PRICE
    583.73€
    144.83€ / Pack of 1
    Save 438.90€
    75% Off
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Material PTFE
Overall Length (Metric) 74 mm
For Use With (Application) For distributing liquids or gases
Min. Operating Temperature (Metric) -30°C
Thread Size GL18
Max. Operating Temperature (Metric) 150°C
Product Type Three-way Stopcock