Learn More
PTGIS Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27061-1-AP-150UL
 View more versions of this product
Product Code. 18045395
  • 410.00€ / 150µL
Description
The PTGIS antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PTGIS. This antibody recognizes human, mouse antigen. The PTGIS antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
PTGIS
550 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Western Blot, Immunofluorescence, Immunohistochemistry, ELISA
CYP8, CYP8A1, PGIS, Prostacyclin synthase, Prostaglandin I2 synthase, PTGI, PTGIS
150 μL
5740
Store at -20°C. Stable for one year after shipment.