missing translation for 'onlineSavingsMsg'
Learn More

PTP Sample for Elemental Analysis of Lube Oil, VHG™

Product Code. 17941596
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17941596 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17941596 Supplier VHG Labs Supplier No. VHGPTPLUBEMO25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

PTP Sample for Elemental Analysis of Lube Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Barium/Calcium/Chlorine/Magnesium/Molybdenum/Phosphorus/Zinc
CAS 7440-39-3, 7440-70-2, 7782-50-5, 7439-95-4, 7439-98-7, 7723-14-0
Methods XRF/RDE
Analyte or Component Names Barium/Calcium/Chlorine/Magnesium/Molybdenum/Phosphorus/Zinc
Shipping Condition Room Temperature
Certifications/Compliance ISO/IEC 17043
Solvent or Matrix Lubricating Oil
Quantity 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.