missing translation for 'onlineSavingsMsg'
Learn More

PTP Sample for Particle Count Analysis (traceable to NIST SRM 2806a), VHG™

Product Code. 30003075
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003075 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003075 Supplier VHG Labs Supplier No. VHGVPTPPC2806A100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

PTP Sample for Particle Count Analysis (traceable to NIST SRM 2806a)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Particle Count Analysis
Analyte or Component Names Particle Count Analysis
Shipping Condition Room Temperature
Certifications/Compliance ISO/IEC 17043
IUPAC Name Particle Count Analysis
Solvent or Matrix Hydrocarbon Oil
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.