missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN13/PTPL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
203.00€ - 400.00€
Specifications
| Antigen | PTPN13/PTPL1 |
|---|---|
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:500-1:3000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18079893
|
Novus Biologicals
NB100-56139SS |
0.025 mL |
203.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18701294
|
Novus Biologicals
NB100-56139 |
400.00€
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
PTPN13/PTPL1 Polyclonal specifically detects PTPN13/PTPL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| PTPN13/PTPL1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Protein Phosphatase | |
| Neat whole antisera with 0.05% Sodium Azide | |
| hPTPE1, PNP1, protein tyrosine phosphatase, non-receptor type 13 (APO-1/CD95 (Fas)-associatedphosphatase), protein-tyrosine phosphatase 1, Fas-associated, PTP1EDKFZp686J1497, PTP-BASEC 3.1.3.48, PTP-E1, PTPL1nonreceptor type 13, tyrosine-protein phosphatase non-receptor type 13 | |
| PTPN13 | |
| IgG | |
| Unpurified | |
| FAP-1 typically migrates at ∼270 kDa on western blots. Thus for SDS-PAGE and western blot, the researcher should use techniques optimized for the transfer of proteins with high molecular weights. The actual molecular weight of FAP-1 observed depends on the isoform expression. |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:500-1:3000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen 1:10-1:500 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| Q12923 | |
| 5783 | |
| A recombinant protein corresponding to amino acids 1279 to 1883 of human FAP-1 protein was used as immunogen; GenBank no. NP_542414.1. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title