missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN13 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25944-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP is a large intracellular protein. It has a catalytic PTP domain at its C-terminus and two major structural domains: a region with five PDZ domains and a FERM domain that binds to plasma membrane and cytoskeletal elements. This PTP was found to interact with, and dephosphorylate, Fas receptor and IkappaBalpha through the PDZ domains. This suggests it has a role in Fas mediated programmed cell death. This PTP was also shown to interact with GTPase-activating protein, and thus may function as a regulator of Rho signaling pathways. Four alternatively spliced transcript variants, which encode distinct proteins, have been reported.Specifications
| PTPN13 | |
| Polyclonal | |
| Unconjugated | |
| PTPN13 | |
| FAP 1, hPTPE1, PNP1, PTP BAS, PTP BL, PTP E1, PTP1E, PTPL1, PTPLE, PTPN13 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5783 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q12923 | |
| PTPN13 | |
| PTPN13 Fusion Protein Ag22854 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction