missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTPN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579895
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: HELA whole cell, JURKAT whole cell. IHC: rat kidney tissue, human intestinal cancer tissue.
T-cell protein tyrosine phosphatase (TC-PTP), also known as PTPT and PTPN2, is an enzyme that removes phosphate groups covalently attached to tyrosine residues in proteins. This enzyme has two C-terminal end splice variants with distinctly different subcellular localizations. The shorter 45 kilodalton isoform is exclusively nuclear in resting cells, but redistrubutes to the cytosol upon stimulation with growth factors and cellular stress. The longer 48 kilodalton isoform is exclusively found in the endoplasmic reticulum 3 and seems to have distinctly different physiologic substrates from the smaller isoform. Although found in many cell types and tissues, TC-PTP is particularly prominent in hemopoietic cell types. Knockout mice lacking TC-PTP are born viable but die 3 to 5 weeks after birth of erythropoietic and lymphopoietic deficits 7, indicating a critical role for TC-PTP in bone marrow maturation. TC-PTP will dephosphorylate a wide range of phosphoproteins, such as p52 Shc 6 and receptors for EGF 1, Insulin 8 and growth hormone.
Specifications
| PTPN2 | |
| Polyclonal | |
| Unconjugated | |
| PTPN2 | |
| AI325124; hypothetical protein LOC540888; MPTP; protein tyrosine phosphatase non-receptor type 2; protein tyrosine phosphatase, non-receptor type 2; protein-tyrosine phosphatase PTP-2; Protein-tyrosine phosphatase PTP-S; PTN2; PTPN 2; PTPN2; Ptpt; T cell protein-tyrosine phosphatase; TC PTP; T-cell protein tyrosine phosphatase; T-cell protein-tyrosine phosphatase; TCELLPTP; TCPTP; TC-PTP; tyrosine-phosphatase; Tyrosine-protein phosphatase non-receptor type 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 117063, 5771 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P17706, P35233 | |
| PTPN2 | |
| E.coli-derived human TCPTP recombinant protein (Position: M1-C330). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction