missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN3 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18412H
This item is not returnable.
View return policy
Description
PTPN3 Polyclonal antibody specifically detects PTPN3 in Human samples. It is validated for Western Blot
Specifications
| PTPN3 | |
| Polyclonal | |
| Western Blot | |
| DKFZp686N0569, EC 3.1.3.48, protein tyrosin phosphatase H1, protein tyrosine phosphatase, non-receptor type 3, Protein-tyrosine phosphatase H1, PTP-H1, PTPH1cytoskeletal-associated protein tyrosine phosphatase, tyrosine phosphatase, tyrosine-protein phosphatase non-receptor type 3 | |
| Recognizes region between residues Between between 175 and 225.. | |
| 0.1 mL | |
| Immunology, Signal Transduction | |
| 5774 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction