Learn More
Invitrogen™ PTPRF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552973
Description
Immunogen sequence: WHPPKELPGE LLGYRLQYCR ADEARPNTID FGKDDQHFTV TGLHKGTTYI FRLAAKNRAG LGEEFEKEIR TPEDLPSGFP QNLHVTGLTT STTELAWDPP VLAERNGRII SYTVVFRDIN SQQE Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.
Possible cell adhesion receptor. It possesses an intrinsic protein tyrosine phosphatase activity (PTPase).The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate.
Specifications
| PTPRF | |
| Polyclonal | |
| Unconjugated | |
| PTPRF | |
| AA591035; BNAH2; FLJ43335; FLJ45062; FLJ45567; LAR; LAR protein; LARFN5C; LARS; LCA homolog; LCA-homolog; leukocyte antigen-related (LAR) PTP receptor; leukocyte antigen-related tyrosine phosphatase; leukocyte common antigen related; protein tyrosine phosphatase receptor-type F; protein tyrosine phosphatase, receptor type F; protein tyrosine phosphatase, receptor type, F; protein tyrosine phosphatase, receptor type, F polypeptide; Ptprf; PTPRF protein; receptor-linked protein-tyrosine phosphatase LAR; receptor-type tyrosine-protein phosphatase F; RPTP-LAR | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5792 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P10586 | |
| PTPRF | |
| Recombinant protein corresponding to Human PTPRF. Recombinant protein control fragment (Product #RP-90310). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.