missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PU.1/Spi-1 Antibody (732322) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5870Y
This item is not returnable.
View return policy
Description
PU.1/Spi-1 Monoclonal antibody specifically detects PU.1/Spi-1 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| PU.1/Spi-1 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 732322 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| hematopoietic transcription factor PU.1, OF, PU.1, SFPI131 kDa transforming protein, SPI-1, SPI-1 proto-oncogene, SPI-A, spleen focus forming virus (SFFV) proviral integration oncogene spi1,31 kDa-transforming protein, transcription factor PU.1 | |
| E. coli-derived recombinant human PU.1/Spi-1, Met1-Lys169, Accession # NP_001074016 | |
| 0.1 mL | |
| Innate Immunity | |
| 6688 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction