missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Pump Bottle with Dispenser

Product Code. 17966272 Shop All Bürkle Products
Change view
Click to view available options
Capacity (English):
8.8 oz.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (English) Unit Size
17966272 8.8 oz. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17966272 Supplier Buerkle™ Supplier No. 03097250
  • ONLINE EXCLUSIVE PRICE
    1.50€
    1.28€ / Pack of 1
    Save 0.22€
    14% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The pump bottle with dispenser allows precise and hygienic dosing of various liquids. It protects against contamination, fits comfortably in the hand and shows the contents at a glance. The pump head can be securely locked in place.

The pump bottle with dispenser delivers an accurate, clean and dosed amount of the medium. It is excellent for a variety of liquids such as reagents, detergents, disinfectants or lotions.

The dispenser pump protects the contents from contamination and allows hygienic dispensing as the pump minimises air and germ ingress and prevents fingers or other objects from coming into contact with the contents.

The crystal clear empty bottle fits comfortably in the hand and allows the contents to be clearly seen. It can be used multiple times and provides efficient residue emptying.

The pump head of the dispenser pump can be locked in place with a twist-and-lock cap, ensuring that no liquid can escape.

  • Dispenses a precise amount
  • Protects the contents
  • Tightly closes with extra long closure thread
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
Color Transparent
Material Polypropylene
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Autoclavable Not Autoclavable
Capacity (English) 8.8 oz.
Shape High Shoulder
Disposable Reusable
Product Type Dosing Pump
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.