missing translation for 'onlineSavingsMsg'
Learn More

Pure Deionised Water, Waternation

Product Code. 30130295 Shop All Waternation Medical Division Products
Change view
Click to view available options
Quantity:
10 x 100 mL
10 x 500 mL
6 x 1000 mL
Unit Size:
1 L
5 L
6 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30130295 10 x 500 mL 5 L
30130305 10 x 100 mL 1 L
30130301 6 x 1000 mL 6 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30130295 Supplier Waternation Supplier No. MLG5500/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

EP grade Pure Deionised Water. Complies with the requirements of ASTM D1193-06, ISO 3696 Type 2A and 3A.

  • Ready to use
  • No chemical additives
  • Batch traceability
  • Certificate of conformity and analysis
  • Reduce expensive test fails due to impurities
  • High performance
  • High accuracy
  • Suppress water-forming side reactions
  • Provide consistent performance across many sample types

The MLG range, a comprehensive suite of five high grade pure waters, in assorted sizes, ranging from simple deionised to molecular biology grades, catering to the majority of pure water lab uses.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

pH 7
Conductivity 1 μS/cm
Grade EP
Packaging PET Plastic Bottle
Certificate of Analysis Yes
Quantity 10 x 500 mL
For Use With (Application) Glassware Washing and Rinsing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.