missing translation for 'onlineSavingsMsg'
Learn More

Veolia Water Systems™ Purification Cartridge

Product Code. 11950457
Change view
Click to view available options
For Use With (Equipment):
For PURELAB Classic Mk2
For PURELAB Option-Q
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11950457 For PURELAB Classic Mk2 Each
11970427 For PURELAB Option-Q Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11950457 Supplier Veolia Water Systems™ Supplier No. LC186

Please to purchase this item. Need a web account? Register with us today!

Edge

Reliable, robust, easy-to-use general water purification cartridge and filter kits ideal for removing ionic contaminants to achieved specific water quality.

  • General water purification cartridge and filter kits
  • Purification cartridge
  • Removes ionic contaminants to achieve specified water quality
  • Compatible with PURELAB Classic Mk2

Specifications

For Use With (Application) Removes ionic contaminants to achieve specified water quality
For Use With (Equipment) For PURELAB Classic Mk2
Quantity 1 Each
Type Cartridge
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.