Learn More
Euro Protection™ Purofort™ Safety Boots
Product Code. 15849226
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15849226 37 Each
15859226 38 Each
15869226 39 Each
15879226 40 Each
15889226 41 Each
15899226 42 Each
15809236 43 Each
15819236 44 Each
15829236 45 Each
15839236 46 Each
15849236 47 Each
15859236 48 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15849226 Supplier Euro Protection™ Supplier No. 9PUSG37
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Euro Protection™ Purofort™ Safety Boots provide flexible and lightweight protection and cold insulation and feature a steel toe cap for protection.
  • Flexible and lightweight for use all day long
  • Cold insulation
  • Tear and wear resistant
  • Purofort™ upper
  • Polyamide lining
  • Polyurethane insole
  • Steel toe cap
  • Steel, puncture-resistant sole
  • Purofort™ outsole
  • Suitable for use in farming industry
Specifications
Color Green
For Use With (Application) Agriculture/Farming
Liner Material Polyamide
Size 37
Sole Features Polyurethane insole, steel, puncture-resistant sole, Purofort™ outsole.
Sole Material Purofort™
Toe Material Steel
Weight (Metric) 900 g per boot in size 9.5
Certifications/Compliance (Europe) EN ISO 20345 : 2011
Product Type Safety Boot