Learn More
Puromycin Dihydrochloride, Fisher BioReagents™
Product Code. 10054207
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10054207 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 10054207 Supplier Fisher BioReagents Supplier No. BP2956100
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Inhibits protein synthesis | CAS: 58-58-2 | C22H31Cl2N7O5 | 544.43 g/mol
Inhibits protein synthesis
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Puromycin Dihydrochloride
Type Puromycin Dihydrochloride
Physical Form Solid
CAS 58-58-2
Melting Point 168°C
Source Streptomyces alboniger
Packaging Clear Glass
Water <12.0
Color Beige
Quantity 100 mg
Assay Percent Range > 98 %
Molecular Formula C22H31Cl2N7O5
MDL Number MFCD00150080
Merck Index 15, 8054
InChI Key MKSVFGKWZLUTTO-MLYJQVMKNA-N
SMILES Cl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1
IUPAC Name (2S)-2-amino-N-[(2S,3S,4S,5R)-5-[6-(dimethylamino)purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide;dihydrochloride
Molecular Weight (g/mol) 544.43
PubChem CID 131632508
Formula Weight 544.44
Percent Purity 98% min.
Show More Show Less
compliance-icons
Product Identifier
  • Puromycin dihydrochloride hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.