Learn More
Euro Protection™ PVC Acifort Boot
Product Code. 12014435
Change view
Click to view available options
Size: 42
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12014435 Size: 42
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12014435 Supplier Euro Protection™ Supplier No. 9ACSA42
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Specifications
Color Black
For Use With (Application) Dangerous and/or wet places, Handling of heavy objects, Industry, Agriculture/horticulture
Liner Material Polyester
Material PVC
Size 42
Sole Features Steel midsole
Sole Material PVC
Toe Material Steel
Waterproof Yes