missing translation for 'onlineSavingsMsg'
Learn More

Advantec Mfs™ PVC Vacuum Manifold

Product Code. 11721953
Change view
Click to view available options
:
Dimensions (L x W x H): 87.6 x 12 x 17.8cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11721953 Dimensions (L x W x H): 87.6 x 12 x 17.8cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11721953 Supplier Advantec Mfs™ Supplier No. 313600
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Lightweight, standard cup, multi-station vacuum manifold. Advantec Mfs™ PVC Vacuum Manifold is sterilizable with ethanol or formaldehyde.

  • Lightweight PVC construction
  • Standard cup branches
  • Sterlizable with ethanol or formaldehyde
  • PTFE stopcocks

Specifications

Material PVC
Manifold Type Filtration Manifold
Quantity 1Pack
Length (Metric) 880 mm
Width (Metric) 200 mm
Height (Metric) 200 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.