Learn More
Euro Protection™ PVC Gloves
Product Code. 12078085
Change view
Click to view available options
Size:
8
9
10
Color:
Green
Red
Length (Metric):
27 cm
36 cm
40 cm
Unit Size:
10 pairs
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Color Length (Metric) unitSize
12078085 8 Red 36 cm 10 pairs
12088085 9 Red 36 cm 10 pairs
12018085 9 Red 27 cm 10 pairs
12008095 10 Red 40 cm 10 pairs
12058095 9 Green 27 cm 10 pairs
12078095 8 Green 40 cm 10 pairs
12088095 9 Green 40 cm 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12078085 Supplier Euro Protection™ Supplier No. 3634
 Restricted Item
Estimated Shipment: 15-10-2026
Specifications
Certifications/Compliance CE
Color Red
Material PVC
Product Type Gloves
Size 8
Lining Cotton
Quantity 10Pack
Length (Metric) 36 cm