Learn More
Dostmann Electronic™ PVC Immersion Probes
Product Code. 11862881
Change view
Click to view available options
Length (Metric):
150 mm
300 mm
Description:
3 mm dia.
3mm dia.
6mm dia.
Sensor Types:
Class 1/10
Class 1/3
Class B
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Description Sensor Types unitSize
11862881 300 mm 6mm dia. Class 1/10 Each
10259461 300 mm 3 mm dia. Class B Each
10688002 150 mm 3 mm dia. Class B Each
10331492 150 mm 3mm dia. Class 1/3 Each
11852881 300 mm 3mm dia. Class 1/10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Récupel-bijdrage uitgesloten
Product Code. 11862881 Supplier Dostmann Electronic™ Supplier No. 60001784
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
With an accuracy of ± 0.3°C, and a temperature range of -200 to + 500°C, this immersion probe is ideal for use in laboratory, scientific, and other research environments.
  • Accessories for hand-held meters Series P700
  • PT100 immersion probe
  • Immersion probe Pt100
  • -50 to +350°C, Cl. B, 150 x 3mm
  • Handheld meter accessory
Specifications
Type Immersion Probe
Description 6mm dia.
Material PVC
Sensor Types Class 1/10
Temperature Range (Metric) -200°C to +450°C
Length (Metric) 300 mm