missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PVRL3/Nectin 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11213-1-AP-150UL
This item is not returnable.
View return policy
Description
Nectins (e.g., PVRL1; MIM 600644) are immunoglobulin-like adhesion molecules that interact with afadin (AF6; MIM 159559). Afadin is an actin filament-binding protein that connects nectins to the actin cytoskeleton. The nectin-afadin system organizes adherens junctions cooperatively with the cadherin (see MIM 192090)-catenin (see MIM 116805) system in epithelial cells.Specifications
| PVRL3/Nectin 3 | |
| Polyclonal | |
| Unconjugated | |
| NECTIN3 | |
| CD113, CDw113, nectin 3, poliovirus receptor related 3, PPR3, PRR3, PVRL3, PVRR3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25945, 288124, 58998 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9JLB9, Q9NQS3 | |
| Nectin3 | |
| PVRL3, Nectin 3 Fusion Protein Ag1712 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction