Learn More
PVRL3/Nectin 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11213-1-AP-20UL
 View more versions of this product
Product Code. 16828143
  • 145.00€ / 20µL
Description
Nectins (e.g., PVRL1; MIM 600644) are immunoglobulin-like adhesion molecules that interact with afadin (AF6; MIM 159559). Afadin is an actin filament-binding protein that connects nectins to the actin cytoskeleton. The nectin-afadin system organizes adherens junctions cooperatively with the cadherin (see MIM 192090)-catenin (see MIM 116805) system in epithelial cells.
Specifications
Specifications
PVRL3/Nectin 3
Polyclonal
Unconjugated
NECTIN3
CD113, CDw113, nectin 3, poliovirus receptor related 3, PPR3, PRR3, PVRL3, PVRR3
Rabbit
Antigen Affinity Chromatography
RUO
25945, 288124, 58998
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9JLB9, Q9NQS3
Nectin3
PVRL3, Nectin 3 Fusion Protein Ag1712
20 μL
Primary
Mouse, Human, Rat