missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PXMP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | PXMP2 |
|---|---|
| Concentration | 0.17 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16803305
|
Proteintech
24801-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16893295
|
Proteintech
24801-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Peroxisomes are subcellular organelles with important functions in lipid metabolism that are found in virtually all eucaryotic cells. The peroxisomal membrane contains a number of integral and peripheral membrane proteins involved in the import of peroxisomal matrix proteins and the transport of metabolites across the membrane. Pxmp2 (Peroxisomal membrane protein 2), also known as PMP22, is a 195 amino acid multi-pass membrane protein that belongs to the peroxisomal membrane protein PXMP2/4 family. The Pxmp2 protein interacts with both Peroxin 19 and Siva proteins. The Peroxin 19 protein binds multiple peroxisomal membrane proteins (PMPs), is predomitly cytoplasmic, and is required for peroxisome membrane synthesis. Pxmp2 is the most abundant peroxisomal membrane protein in higher eukaryotes. Pxmp2′s expression is tissue-specific with highest levels of expression in liver, kidney and heart tissue. It has been suggested that Pxmp2 is involved in pore-forming activity and may contribute to the broad permeability of the peroxisomal membrane. Disruption of the mouse Pxmp2 gene leads to partial restriction of peroxisomal membrane permeability to solutes in vitro and in vivo. The Pxmp2 gene is conserved in chimpanzee, dog, cow, mouse, rat, zebrafish, fruit fly and mosquito, and maps to human chromosome 12q24.33.Specifications
| PXMP2 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| P42925, Q07066, Q9NR77 | |
| 19301, 29533, 5827 | |
| PXMP2 Fusion Protein Ag20590 | |
| Primary | |
| -20°C | |
| PXMP2 |
| 0.17 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Peroxisomal membrane protein 2, PMP22, PXMP2 | |
| PXMP2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title