Learn More
PXN Monoclonal Antibody (OTI6G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811265
Description
Paxillin is a focal adhesion protein and a substrate for several tyrosine kinases such as src, FAK, and p210BRC/ABL. The tyrosine phosphorylation of paxillin is affected by conditions that change cell-cell adhesion. Paxillin associates tightly with FAK and Crk through its SH2 domain. This interaction is independent of the extracellular matrix. Although paxillin was initially discovered in fibroblasts, its phosphorylation may also be important during neurite extension during differentiation.Specifications
| PXN | |
| Monoclonal | |
| Unconjugated | |
| PXN | |
| paxillin | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5829 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI6G9 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P49023 | |
| PXN | |
| Full length human recombit protein of human PXN produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.