Learn More
PXN Monoclonal Antibody (OTI6G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811265S
Description
PXN Monoclonal antibody specifically detects PXN in Human samples. It is validated for Western BlotSpecifications
| PXN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P49023 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5829 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI6G9 | |
| Unconjugated | |
| PXN | |
| adaptor protein; AW108311; AW123232; FLJ16691; hCG_1778014; I79_001229; myocardial ischemic preconditioning associated protein 7; Pax; PAX-1; PAXI; Paxillin; paxillin a; paxillin alpha; pxn; pxna; testicular tissue protein Li 134; TGFB1I1; transforming growth factor beta 1 induced transcript 1; wu:fw71f12 | |
| Full length human recombinant protein of human PXN produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.