Learn More
PXR/NR1I2 Antibody (V11P4G11*E7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50519-0.025ml
 View more versions of this product
Product Code. 18655958
  • 190.00€ / 0.03mL
Description
PXR/NR1I2 Monoclonal antibody specifically detects PXR/NR1I2 in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PXR/NR1I2
Monoclonal
1.1 mg/mL
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10-1:500
nuclear receptor subfamily 1 group I member 2, nuclear receptor subfamily 1, group I, member 2, ONR1, Orphan nuclear receptor PAR1, Orphan nuclear receptor PXR, PAR, PAR1, PAR2, Pregnane X receptor, PRR, PXRPARq, SAR, Steroid and xenobiotic receptor, SXRBXR
Ovalbumin-conjugated synthetic peptide EMRSMNEEYT (C-terminal sequence)
0.025 mL
Stem Cell Markers, Transcription Factors and Regulators
8856
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
V11P4G11*E7
Unconjugated
PBS
Mouse
Protein A purified
RUO
Primary
Zebrafish