missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PYGO2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148735
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The PYGO2 gene encodes pygopus family PHD finger 2, a key component of the Wnt signaling pathway, which is critical for embryonic development, cell proliferation, and differentiation. PYGO2 operates as a chromatin remodeling factor, interacting with the beta-catenin-LEF/TCF transcriptional complex, facilitating the transcription of Wnt target genes by recruiting chromatin-modifying enzymes. This activity is vital for maintaining the balance between stem cell renewal and differentiation in various tissues, including the brain and the skin. Additionally, PYGO2 plays significant roles in spermatogenesis and hematopoiesis by modulating gene expression necessary for these processes. Dysregulation of PYGO2 expression can lead to developmental abnormalities and is often implicated in cancer progression, indicating its potential as a biomarker for tumorigenic stem cell-like properties and metastatic behavior. Research continues to explore PYGO2's mechanisms and interactions to understand its contributions to development and its implications in disease states, particularly in cancer biology.
Specifications
| PYGO2 | |
| Polyclonal | |
| Unconjugated | |
| PYGO2 | |
| 1190004M21Rik; mpygo2; PP7910; PYGO2; pygopus 2; pygopus family PHD finger 2; pygopus homolog 2; RGD1308535 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 90780 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9BRQ0 | |
| PYGO2 | |
| E. coli-derived recombinant human Pygopus-2, Met160-Glu336. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction