Learn More
Pyrazine-2,3-dicarboxylic acid, 98%
Product Code. 11479617
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479617 100 g 100g
11459617 5 g 5g
11469617 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11479617 Supplier Thermo Scientific Alfa Aesar Supplier No. B22108.22
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 89-01-0 | C6H4N2O4 | 168.108 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 89-01-0
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.108
MDL Number MFCD00006131
InChI Key ZUCRGHABDDWQPY-UHFFFAOYSA-N
Synonym 2,3-pyrazinedicarboxylic acid, pyrazine-2,3-dicarboxylate, 2,3-dicarboxypyrazine, 2,3-pyrazine dicarboxylic acid, unii-qnn35wof29, 2,3-pyrazinedicarboxylicacid, qnn35wof29, 2,3-pyrazinedcarboxylicacid, pubchem8610, acmc-209qye
PubChem CID 66628
IUPAC Name pyrazine-2,3-dicarboxylic acid
SMILES C1=CN=C(C(=N1)C(=O)O)C(=O)O
Specifications
Melting Point ∼188°C (decomposition)
Quantity 100 g
Beilstein 147982
Merck Index 14,7958
Formula Weight 168.11
Percent Purity 98%
Chemical Name or Material Pyrazine-2,3-dicarboxylic acid