Learn More
Pyrex™ Borosilicate Glass Kjeldahl Flask
Product Code. 12555017
Change view
Click to view available options
Capacity (Metric):
150 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12555017 150 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12555017 Supplier Pyrex Supplier No. S/3L2985
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Specifications
Certifications/Compliance NF B 35004
Capacity (Metric) 150 mL
Material Borosilicate Glass
Height (Metric) 275 mm
Bottom Shape Round
Diameter (Metric) Outer 66 mm