Learn More
Pyrex™ Borosilicate Glass Spare Glass Keys
Product Code. 15633076
Change view
Click to view available options
Diameter (Metric):
12.5 mm
14.5 mm
Plug Bore:
1.5 mm
2.5 mm
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Plug Bore Unit Size
15633076 14.5 mm 2.5 mm Pack of 5
12454761 12.5 mm 1.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15633076 Supplier Pyrex Supplier No. SST103K
  • ONLINE EXCLUSIVE PRICE
    109.40€
    87.74€ / Pack of 5
    Save 21.66€
    19% Off
Estimated Shipment: 12-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Manufactured from borosilicate glass
  • Replacement keys for SSD: series stopcocks
  • Manufactured from borosilicate glass
  • Supplied complete with key retaining device
Specifications
Type Spare Glass Key
Plug Bore 2.5 mm
Diameter (Metric) 14.5 mm
Material Borosilicate Glass