Learn More
Pyrex™ Graduated Dropping Funnel with Glass Key
Product Code. 12433481
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12433481 50 mL Each
11333684 1000 mL Each
11313684 250 mL Each
11323684 500 mL Each
11303684 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12433481 Supplier Pyrex Supplier No. 3590/01
  • ONLINE EXCLUSIVE PRICE
    144.90€
    119.82€ / Pack of 1
    Save 25.08€
    17% Off
Estimated Shipment: 08-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Used to transfer fluids.
  • Complies with ISO 4800 type 4
  • Manufactured from Pyrex borosilicate glass
  • Double graduated scale showing approximate volume contained/dispensed
  • Graduations in white enamel
  • With interchangeable glass key
  • Supplied with glass stopper
Specifications
Capacity (Metric) 50 mL
Material Borosilicate Glass
Graduated Yes
Graduations 1mL
Product Type Dropping Funnel