missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2,4-dicarboxylic acid, 98%, may cont. up to ca 10% water, Thermo Scientific™

Product Code. 11497027
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497027 100 g 100g
11477027 5 g 5g
11487027 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497027 Supplier Thermo Scientific Alfa Aesar Supplier No. A14409.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an in vitro and in cell inhibitor, as well as a known inhibitor of the histone lysine demethylases. 2,4-Pyridinedicarboxylic acid has been used in a study to determine that ruthenium(II) complexes exert antimetastatic effects on several tumor cell lines in vitro, achieved mostly by the effect on cell adhesion, migration and angiogenesis. It blocks the activity of 2-OG oxygenases, which include certain lysine demethylases and a variety of hydroxylases (e.g., prolyl, collagen, lysyl). 2,3 2,4-PDCA inhibits several Jumonji domain-containing lysine demethylases when used at low micromolar concentrations. Through its effects on hydroxylases, including prolyl hydroxlase 1 (IC50 = 1.5 µM), It modulates hypoxia inducing factor turnover, collagen synthesis, and plant cell wall formation. It can inhibit zinc-dependent enzymes, like metallo-β-lactamase.8 2,4-PDCA also affects and is translocated by organic anion transporters.

Solubility
Soluble in water at 20°C 4.5 g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 499-80-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00149413,MFCD00006296
InChI Key MJIVRKPEXXHNJT-UHFFFAOYSA-N
Synonym 2,4-pyridinedicarboxylic acid, lutidinic acid, 2,4-lutidinic acid, 2,4-pyridinedicarboxylicacid, hydrate 1:1, 4nrq, pd2, 4-carboxypicolinic acid, pubchem16314, pyridine carboxylate, 6a, acmc-1bnb9
PubChem CID 10365
ChEBI CHEBI:44737
IUPAC Name pyridine-2,4-dicarboxylic acid
SMILES OC(=O)C1=CC=NC(=C1)C(O)=O

Specifications

Melting Point ∼245°C (decomposition)
pH 2.5 to 3.5
Quantity 100 g
Beilstein 131631
Solubility Information Soluble in water at 20°C 4.5g/L.
Formula Weight 167.12
Percent Purity 98%
Assay may cont. up to ≈10% water
Chemical Name or Material Pyridine-2,4-dicarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.