missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2-carboxamide, 98%

Product Code. 11478386
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11478386 25 g 25g
11468386 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11478386

Brand: Thermo Scientific Alfa Aesar A15410.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1452-77-3 | C6H6N2O | 122.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1452-77-3
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
MDL Number MFCD00023483
InChI Key IBBMAWULFFBRKK-UHFFFAOYSA-N
Synonym picolinamide, 2-pyridinecarboxamide, picolinoylamide, 2-carbamoylpyridine, picolinic acid amide, 2-picolinamide, 2-aminocarbonyl-pyridine, alpha-picolinamide, pyridinecarboxamide, unii-i3550ccl59
PubChem CID 15070
ChEBI CHEBI:8200
IUPAC Name pyridine-2-carboxamide
SMILES NC(=O)C1=CC=CC=N1

Specifications

Melting Point 104°C to 108°C
Boiling Point 143°C (20 mmHg)
Quantity 25 g
Beilstein 109617
Formula Weight 122.13
Percent Purity 98%
Chemical Name or Material Pyridine-2-carboxamide
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.