missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-4-carbonitrile, Mikromol™

Product Code. 17951366
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17951366 100 mg 100mg
17990746 25 mg 25mg
30463762 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17951366 Supplier Mikromol Supplier No. MM0845.03
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pyridine-4-carbonitrile
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyridine-4-carbonitrile
CAS 100-48-1
Analyte or Component Names Pyridine-4-carbonitrile
Shipping Condition Room Temperature
Molecular Formula C6 H4 N2
Certifications/Compliance ISO 9001
Synonym 4-Pyridinecarbonitrile, Isonicotinonitrile (6CI,8CI), 4-Azabenzonitrile, 4-Cyanopyridine, 4-Pyridinenitrile, 4-Pyridyl cyanide, 4-Pyridylcarbonitrile, Isonicotinic acid nitrile, NSC 60681, gamma-Cyanopyridine, Fampridine Impurity 2, Ethionamide Impurity B, Isoniazid Impurity C
InChI Formula InChI=1S/C6H4N2/c7-5-6-1-3-8-4-2-6/h1-4H
SMILES N#Cc1ccncc1
IUPAC Name pyridine-4-carbonitrile
Molecular Weight (g/mol) 104.11
Quantity 100 mg
Formula Weight 104.0374 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.