missing translation for 'onlineSavingsMsg'
Learn More

Pyridostigmine Bromide, Mikromol™

Product Code. 18051100
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18051100 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18051100 Supplier Mikromol Supplier No. MM0429.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pyridostigmine Bromide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyridostigmine Bromide
CAS 101-26-8
Analyte or Component Names Pyridostigmine Bromide
Shipping Condition Room Temperature
Molecular Formula C9 H13 N2 O2 . Br
Certifications/Compliance ISO 17034
Synonym Pyridostigmine Bromide, 3-Hydroxy-1-methylpyridinium bromide dimethylcarbamate, Dimethylcarbamic acid ester with 3-hydroxy-1-methylpyridinium bromide, 1-Methyl-3-hydroxypyridinium bromide dimethylcarbamate, 3-(Dimethylcarbamoyloxy)-1-methylpyridinium bromide, 3-(Dimethylcarbamyloxy)-1-methylpyridinium bromide, 3-[[(Dimethylamino)carbonyl]oxy]-1-methylpyridinium bromide, Kalimin, Kalymin, Mestinon, Mestinon bromide, NSC 679759, Piridostigmin, Piridostigmin bromide, Pyridostigmine bromide, Regonol, Ro 1-5130
InChI Formula InChI=1S/C9H13N2O2.BrH/c1-10(2)9(12)13-8-5-4-6-11(3)7-8;/h4-7H,1-3H3;1H/q+1;/p-1
SMILES [Br-].CN(C)C(=O)Oc1ccc[n+](C)c1
IUPAC Name (1-methylpyridin-1-ium-3-yl) N,N-dimethylcarbamate;bromide
Molecular Weight (g/mol) 261.12
Quantity 100 mg
Formula Weight 260.016 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.