missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridostigmine-d3 Bromide (1-methyl-d3), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Pyridostigmine-D3 (1-methyl-D3) Bromide |
| Molecular Formula | C9 D3 H10 N2 O2 . Br |
| Quantity | 0.01 g |
| Synonym | 3-[[(Dimethylamino)carbonyl]oxy]-1-(methyl-d3)pyridinium bromide, Pyridostigmine-d3 (1-methyl-d3) bromide, Pyridostigmine-d3 bromide, 3-Hydroxy-1-(methyl-d3)pyridinium dimethylcarbamate bromide, 3-[(Dimethylcarbamoyl)oxy]-1-(methyl-d3)pyridin-1-ium bromide, Pyridostigmine-D3 (1-methyl-D3) bromide |
| InChI Formula | InChI=1 S/C9H13N2O2.BrH/c1-10(2)9(12)13-8-5-4-6-11(3)7-8;/h4-7 H,1-3H3;1 H/q+1;/p-1/i3D3; |
| SMILES | [Br-].[2 H]C([2 H])([2 H])[n+]1 cccc(OC(=O)N(C)C)c1 |
| IUPAC Name | [1-(trideuteriomethyl)pyridin-1-ium-3-yl] N,N-dimethylcarbamate;bromide |
| Molecular Weight (g/mol) | 264.134 |
| Formula Weight | 263.035 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction