missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxine EP Impurity A as Hydrochloride, Mikromol™

Product Code. 17914366
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17914366 25 mg 25mg
17953776 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17914366 Supplier Mikromol Supplier No. MM0021.010025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pyridoxine EP Impurity A as Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol Hydrochloride
CAS 1006-21-9
Analyte or Component Names 6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C8 H9 N O2 . Cl H
Certifications/Compliance ISO 17025
Synonym Pyridoxine Hydrochloride Imp. A (EP), Pyridoxine Imp. A (EP), 6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol Hydrochloride, Pyridoxine Hydrochloride Imp. A (EP) as Hydrochloride, Pyridoxine Hydrochloride Impurity A as Hydrochloride, Pyridoxine Impurity A as Hydrochloride
InChI Formula InChI=1S/C8H9NO2.ClH/c1-5-8(10)7-4-11-3-6(7)2-9-5;/h2,10H,3-4H2,1H3;1H
SMILES Cl.Cc1ncc2COCc2c1O
IUPAC Name 6-methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol;hydrochloride
Molecular Weight (g/mol) 187.62
Quantity 25 mg
Formula Weight 187.04 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.