missing translation for 'onlineSavingsMsg'
Learn More

Pyrifenox, TRC

Product Code. 17982995
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17982995 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17982995 Supplier Toronto Research Chemicals Supplier No. TRCP9988555MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 88283-41-4
Quantity 5 mg
Molecular Formula C14 H12 Cl2 N2 O
Synonym 1-(2,4-Dichlorophenyl)-2-(3-pyridinyl)ethanone O-methyloxime, ACR 3453A, Corado, Dorado, Dorado (pesticide), FD 4060, Korado, Pyrifenox, Ro 15-1297
InChI Formula InChI=1S/C14H12Cl2N2O/c1-19-18-14(7-10-3-2-6-17-9-10)12-5-4-11(15)8-13(12)16/h2-6,8-9H,7H2,1H3
SMILES CON=C(Cc1cccnc1)c2ccc(Cl)cc2Cl
IUPAC Name 1-(2,4-dichlorophenyl)-N-methoxy-2-pyridin-3-ylethanimine
Molecular Weight (g/mol) 295.16
Formula Weight 294.0327
Chemical Name or Material Pyrifenox
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.