missing translation for 'onlineSavingsMsg'
Learn More

Pyrimethamine, TRC

Product Code. 30135934
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135934 50 mg 50mg
30133005 100 mg 100mg
18003930 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135934 Supplier Toronto Research Chemicals Supplier No. TRCP99724050MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-14-0
Molecular Formula C12 H13 Cl N4
Molecular Weight (g/mol) 248.71
Synonym Pyrimethamine, 5-(4-chlorophenyl)-6-ethylpyrimidine-2,4-diamine, 2,4-Diamino-5-(4-chlorophenyl)-6-ethylpyrimidine, 2,4-Diamino-5-p-chlorophenyl-6-ethylpyrimidine, 4753 R.P., 5-(4-Chlorophenyl)-2,4-diamino-6-ethylpyrimidine, 5-(P-Chlorophenyl)-6-ethyl-2,4-diaminopyrimidine, BW 50-63, Chloridin, Chloridine, Darachlor, Darapram, Daraprim, Daraprime, Diaminopyritamin, Erbaprelina, Khloridin, Malacid, Malocid, Malocide, NSC 3061, Pirimecidan, Pirimetamin, Pyrimethamin, Pyrimethamine, RP 4753, Tindurin, Tinduring, WR 2978
IUPAC Name 5-(4-chlorophenyl)-6-ethylpyrimidine-2,4-diamine
SMILES CCc1nc(N)nc(N)c1c2ccc(Cl)cc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H13ClN4/c1-2-9-10(11(14)17-12(15)16-9)7-3-5-8(13)6-4-7/h3-6H,2H2,1H3,(H4,14,15,16,17)
Formula Weight 248.0829
Chemical Name or Material Pyrimethamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.