missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-2-carboxylic acid, 97%

Product Code. 15470948
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15470948 1 g 1g
15490948 250 mg 250mg
15480948 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15470948 Supplier Thermo Scientific Alfa Aesar Supplier No. H54745.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 31519-62-7 | C5H4N2O2 | 124.10 g/mol

2-carboxy-Pyrimidine is a synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-carboxy-Pyrimidine is a synthetic intermediate.

Solubility
Soluble in ethanol, Dimethyl sulfoxide (DMSO), Dimethylformamide (DMF), and solution of Phosphate Buffered Saline (PBS).

Notes
Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31519-62-7
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.10
MDL Number MFCD00856161
InChI Key ZFCHNZDUMIOWFV-UHFFFAOYSA-N
Synonym 2-pyrimidinecarboxylic acid, 2-carboxypyrimidine, pyrimidine carboxylic acid, pyrimidinecarboxylic acid, 2-carboxy-1,3-diazine, 2-pyrimidinecarboxylic acid 6ci,8ci,9ci, 2-pyrimidinecarboxylicacid, 2-carboxy pyrimidine, pubchem9603, pyrimidine-carboxylic acid
PubChem CID 12626245
IUPAC Name pyrimidine-2-carboxylic acid
SMILES OC(=O)C1=NC=CC=N1

Specifications

Quantity 1 g
Solubility Information Soluble in ethanol,Dimethyl sulfoxide (DMSO),Dimethylformamide (DMF),and solution of Phosphate Buffered Saline (PBS).
Formula Weight 124.1
Percent Purity 97%
Chemical Name or Material Pyrimidine-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.