missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyruvate Dehydrogenase E1-alpha subunit, p Ser293 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 43 publications
Brand: Novus Biologicals NB110-93479
This item is not returnable.
View return policy
Description
Pyruvate Dehydrogenase E1-alpha subunit Polyclonal specifically detects Pyruvate Dehydrogenase E1-alpha subunit in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunoblotting, Knockout Validated.
Specifications
| Pyruvate Dehydrogenase E1-alpha subunit (p Ser293) | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.4) with 0.1% Sodium Azide | |
| EC 1.2.4.1, PDHA, PDHE1-A type I, PHE1APDHCE1A, pyruvate dehydrogenase (lipoamide) alpha 1, pyruvate dehydrogenase complex, E1-alpha polypeptide 1, pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial | |
| Rabbit | |
| 43 kDa | |
| 0.05 mL | |
| Cancer, Core ESC Like Genes, Lipid and Metabolism, Signal Transduction, Stem Cell Markers | |
| 5160 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoblot, Immunoassay | |
| 1 mg/mL | |
| Western Blot 1:1000-1:5000, Flow Cytometry reported in scientific literature (PMID 31900478), Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunoprecipitation reported in scientific literature (PMID 27450723), Immunoblotting, Knockout Validated reported in scientific literature (PMID 31742248) | |
| P08559 | |
| PDHA1 | |
| A synthetic peptide surrounding the phosphorylated serine 293 of the human Pyruvate Dehydrogenase E1-alpha subunit protein. [Swiss-Prot: #P08559] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This is specific for the phosphorylated Serine 293 form of the PDHE1 alpha protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction