Learn More
Description
Includes
Q-BioTaq DNA polymerase, high purity dNTPs Mix (>99%) at 10mM each, 10X Q-BioTaq Buffer with Mg, 10X Q-BioTaq Buffer without Mg, 25mM MgCl2 solution
Q-BioTaq DNA Polymerase is a thermostable DNA polymerase which can be used effectively for PCR and DNA sequencing by the chain termination method. This enzyme is encoded by a modified form of the Thermus aquaticus DNA polymerase gene with an N-terminal deletion. The properties of Q-BioTaq DNA Polymerase include high thermostability (98°C) and absence of 5'-3' exonuclease activity. This polymerase is particularly useful in applications where multiple amplifications are required. The enzyme is highly purified and is free of endo- and exonucleases.
Unit Definition: One unit is the amount of enzyme required to catalyse the incorporation of 10nmol of nucleotides into acid insoluble material in 30 min. at 74 °C under assay conditions.
Certifications
Free of RNases and endo- and exonucleases
Specifications
Specifications
| Concentration | 5 U/μL |
| Content And Storage | Q-BioTaq DNA polymerase, high purity dNTPs Mix (>99%) at 10mM each, 10X Q-BioTaq Buffer with Mg, 10X Q-BioTaq Buffer without Mg, 25mM MgCl2 solution. Store at -20°C |
| Polymerase | Q-BioTaq DNA polymerase |
| Quantity | 250U |
| For Use With (Application) | Standard PCR |
Research Use Only (RUO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.